Learn More
Apexbio Technology LLC Ro3280, 50mg. Cas: 1062243-51-9 MFCD: MFCD22665717. PLK1 inhibitor,potent and highly selective
SDP
Catalog No. 501905179
Encompass_Preferred
Ro3280 is a selective inhibitor of PLK1 with IC50 value of 3 nM [1]. Polo-like kinase 1 (PLK1) is an enzyme and plays an important role in regulating cell cycles as an early trigger for G2/M phase transition. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5179 Supplier Apexbio Technology LLC Supplier No. A868150
Ro3280 is a selective inhibitor of PLK1 with IC50 value of 3 nM [1]. Polo-like kinase 1 (PLK1) is an enzyme and plays an important role in regulating cell cycles as an early trigger for G2/M phase transition. Other sizes are available. Please inqury us for quote.