Learn More
Medchemexpress LLC HY-11102 10mg Medchemexpress, RO4929097 CAS:847925-91-1 Purity:>98%
SDP
Catalog No. 502028807
Encompass_Preferred
Medchemexpress, HY-11102 10mg RO4929097 CAS:847925-91-1 RO4929097 (RG-4733) is a γ secretase inhibitor with IC50 of 4 nM, inhibiting cellular processing of Aβ40 and Notch with EC50 of 14 nM and 5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8807 Supplier Medchemexpress LLC Supplier No. HY1110210MG
Medchemexpress, HY-11102 10mg RO4929097 CAS:847925-91-1 RO4929097 (RG-4733) is a γ secretase inhibitor with IC50 of 4 nM, inhibiting cellular processing of Aβ40 and Notch with EC50 of 14 nM and 5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.