Learn More
Apexbio Technology LLC RO4929097(Synonyms: RO-4929097, RO 4929097, Gamma-Secretase Inhibitor RO4929097, RO4929097 γ-secretase inhibitor), 10mM (in 1mL DMSO), CAS: 847925-91-1.
SDP
Catalog No. 5000061068
Encompass_Preferred
RO4929097 (CAS 847925-91-1) is a potent selective small-molecule inhibitor of -secretase an essential protease in the Notch signaling pathway It inhibits -secretase with high specificity (IC50 4 nM EC50 5 nM) showing minimal inhibitory activity against closely related proteases and over 100-fold selectivity versus a broad spectrum of other proteases By preventing Notch receptor cleavage and subsequent signaling activation RO4929097 suppresses proliferation and tumorigenesis across various tumor models including melanoma breast colorectal pancreatic and lung cancers Clinical studies have assessed RO4929097 alone or combined with other anticancer agents for advanced solid tumors
Catalog No. 50-000-61068 Supplier Apexbio Technology LLC Supplier No. A40055.1
RO4929097 (CAS 847925-91-1) is a potent selective small-molecule inhibitor of -secretase an essential protease in the Notch signaling pathway It inhibits -secretase with high specificity (IC50 4 nM EC50 5 nM) showing minimal inhibitory activity against closely related proteases and over 100-fold selectivity versus a broad spectrum of other proteases By preventing Notch receptor cleavage and subsequent signaling activation RO4929097 suppresses proliferation and tumorigenesis across various tumor models including melanoma breast colorectal pancreatic and lung cancers Clinical studies have assessed RO4929097 alone or combined with other anticancer agents for advanced solid tumors