Learn More
Selleck Chemical LLC RO4929097 10mM/1mL 847925-91-1
SDP
Catalog No. 501361400
Encompass_Preferred
RO4929097 is a ? secretase inhibitor with IC50 of 4 nM in a cell-free assay, inhibiting cellular processing of A?40 and Notch with EC50 of 14 nM and 5 nM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1400 Supplier Selleck Chemical LLC Supplier No. S157510MM/1ML
RO4929097 is a ? secretase inhibitor with IC50 of 4 nM in a cell-free assay, inhibiting cellular processing of A?40 and Notch with EC50 of 14 nM and 5 nM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.