Learn More
Selleck Chemical LLC RO4987655 S0372-5MG
SDP
Catalog No. 5000042216
Encompass_Preferred
RO4987655(RG 7167 CH4987655) is an allosteric MEK inhibitor with IC50 of 5 nM RO4987655 exhibits a significant anti-proliferative and antitumor activity
Catalog No. 50-000-42216 Supplier Selleck Chemical LLC Supplier No. S03725MG
RO4987655(RG 7167 CH4987655) is an allosteric MEK inhibitor with IC50 of 5 nM RO4987655 exhibits a significant anti-proliferative and antitumor activity