Learn More
Medchemexpress LLC Ro5-3335 | 30195-30-3 | MFCD00870704 | 99.5% | 259.69 | C13H10ClN3O | 5 MG
SDP
Catalog No. 5000263456
Encompass_Preferred
Ro5-3335 is a benzodiazepine-derived small molecule that inhibits the RUNX1-CBFβ protein-protein interaction and represses RUNX1/CBFB-dependent transactivation. It is used as a research tool in studies of RUNX1-driven transcriptional regulation and core binding factor (CBF) leukemias. The compound is provided as a solid and is available in solution formats for assay use.
Catalog No. 50-002-63456 Supplier Medchemexpress LLC Supplier No. HY1084705MG
Ro5-3335 is a benzodiazepine-derived small molecule that inhibits the RUNX1-CBFβ protein-protein interaction and represses RUNX1/CBFB-dependent transactivation. It is used as a research tool in studies of RUNX1-driven transcriptional regulation and core binding factor (CBF) leukemias. The compound is provided as a solid and is available in solution formats for assay use.