Learn More
Selleck Chemical LLC RO5126766 (CH5126766) 5mg 946128-88-7
SDP
Supplier:  Selleck Chemical LLC S71705MG
Encompass_Preferred
RO5126766 (CH5126766) is a dual RAF/MEK inhibitor with IC50 of 8.2 nM,19 nM, 56 nM, and 160 nM for BRAF V600E, BRAF, CRAF, and MEK1, respectively. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1201413