Learn More
TARGETMOL CHEMICALS INC RO5263397 10MG
SDP
Catalog No. 502577416
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RO5263397 is a TAAR1 specific agonist with oral activity that has been used in antidepressant studies. It has also been found to act as an inhibitor of the enzyme cyclooxygenase-2 (COX-2) which is involved in the synthesis of prostaglandins. purity: 99%
Catalog No. 50-257-7416 Supplier TARGETMOL CHEMICALS INC Supplier No. T1274510MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RO5263397 is a TAAR1 specific agonist with oral activity that has been used in antidepressant studies. It has also been found to act as an inhibitor of the enzyme cyclooxygenase-2 (COX-2) which is involved in the synthesis of prostaglandins. purity: 99%