Learn More
Medchemexpress LLC Ro60-0175 | 169675-08-5 | 99.5% | 226.68 g/mol | C11H12ClFN2 | 25MG
SDP
Catalog No. 5000236837
Encompass_Preferred
Ro60-0175 is a selective 5-HT2C (serotonin) receptor agonist used in pharmacology and behavioral research to investigate serotonergic modulation and addiction-related behaviors; it has been reported to reduce drug self-administration. The compound is supplied as a solid and is also available as a prepared DMSO solution for in vitro work. Relevant documentation such as a data sheet, COA, SDS, H NMR, and LCMS are provided with the product.
Catalog No. 50-002-36837 Supplier Medchemexpress LLC Supplier No. HY12383825MG
Ro60-0175 is a selective 5-HT2C (serotonin) receptor agonist used in pharmacology and behavioral research to investigate serotonergic modulation and addiction-related behaviors; it has been reported to reduce drug self-administration. The compound is supplied as a solid and is also available as a prepared DMSO solution for in vitro work. Relevant documentation such as a data sheet, COA, SDS, H NMR, and LCMS are provided with the product.