Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro60-0175 | 169675-08-5 | 99.5% | 226.68 g/mol | C11H12ClFN2 | 5MG
SDP

Catalog No. 5000237525
Encompass_Preferred

Ro60-0175 is a potent, selective agonist of the 5-HT2C serotonin receptor used as a research reagent in pharmacology and behavioral and behavioral studies. It is supplied as a high-purity solid and as a 10 mM solution in DMSO, with recommended solvents, in vivo formulation protocols, and storage conditions provided for experimental use. Intended for research use only.

  • Purity 99.52%.
  • Molecular weight 226.68 g/mol.
  • Chemical formula C11H12ClFN2.
  • Available as solid (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and 10 mM solution in DMSO (1 mL).
  • In vitro solubility: DMSO 250 mg/mL (≈1102.87 mM).
  • In vivo formulation protocols include DMSO/PEG300/Tween-80/saline, SBE-β-CD formulation, or corn oil.
  • Storage: solid at 4°C protected from light; solutions -80°C for long-term storage.

Catalog No. 50-002-37525 Supplier Medchemexpress LLC Supplier No. HY1238385MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro60-0175 is a potent, selective agonist of the 5-HT2C serotonin receptor used as a research reagent in pharmacology and behavioral and behavioral studies. It is supplied as a high-purity solid and as a 10 mM solution in DMSO, with recommended solvents, in vivo formulation protocols, and storage conditions provided for experimental use. Intended for research use only.

  • Purity 99.52%.
  • Molecular weight 226.68 g/mol.
  • Chemical formula C11H12ClFN2.
  • Available as solid (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and 10 mM solution in DMSO (1 mL).
  • In vitro solubility: DMSO 250 mg/mL (≈1102.87 mM).
  • In vivo formulation protocols include DMSO/PEG300/Tween-80/saline, SBE-β-CD formulation, or corn oil.
  • Storage: solid at 4°C protected from light; solutions -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.