Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro60-0175 fumarate | 169675-09-6 | MFCD06798313 | 99.5% | 342.75 | C15H16ClFN2O4 | 5 MG
SDP

Catalog No. 5000246293
Encompass_Preferred

Ro60-0175 fumarate is a research-grade small molecule agonist selective for the serotonin 5-HT2C receptor, used in pharmacology and neuroscience studies to probe 5-HT2C-mediated signaling in cellular and in vivo models.

  • Selective 5-HT2C receptor agonist for pharmacology research.
  • Reported purity 99.47% by HPLC.
  • Molecular weight 342.75 g/mol.
  • Chemical formula C15H16ClFN2O4.
  • Available as solid 5 MG and as a 10 mM solution in DMSO (1 ML).
  • Soluble in DMSO for solution preparation and dosing in assays.

Catalog No. 50-002-46293 Supplier Medchemexpress LLC Supplier No. HY1031405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro60-0175 fumarate is a research-grade small molecule agonist selective for the serotonin 5-HT2C receptor, used in pharmacology and neuroscience studies to probe 5-HT2C-mediated signaling in cellular and in vivo models.

  • Selective 5-HT2C receptor agonist for pharmacology research.
  • Reported purity 99.47% by HPLC.
  • Molecular weight 342.75 g/mol.
  • Chemical formula C15H16ClFN2O4.
  • Available as solid 5 MG and as a 10 mM solution in DMSO (1 ML).
  • Soluble in DMSO for solution preparation and dosing in assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.