Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro60-0175 fumarate | 169675-09-6 | MFCD06798313 | 99.5% | 342.75 g/mol | C15H16ClFN2O4 | 1 ML
SDP

Catalog No. 5000246708
Encompass_Preferred

Ro60-0175 fumarate is a selective agonist of the serotonin 5-HT2C receptor used in neuroscience and pharmacology research. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid quantities; reported purity is 99.47%, with molecular formula C15H16ClFN2O4 and molecular weight 342.75 g/mol.

  • Selective 5-HT2C receptor agonist for pharmacology studies.
  • Available as 10 mM solution in DMSO (1 mL) and as solids (5, 10, 50 mg).
  • High purity (~99.5%) suitable for research applications.
  • Soluble in DMSO; in vitro solubility approximately 100 mg/mL.
  • Storage guidance provided for powder and solutions to preserve stability.

Catalog No. 50-002-46708 Supplier Medchemexpress LLC Supplier No. HY10314010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro60-0175 fumarate is a selective agonist of the serotonin 5-HT2C receptor used in neuroscience and pharmacology research. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid quantities; reported purity is 99.47%, with molecular formula C15H16ClFN2O4 and molecular weight 342.75 g/mol.

  • Selective 5-HT2C receptor agonist for pharmacology studies.
  • Available as 10 mM solution in DMSO (1 mL) and as solids (5, 10, 50 mg).
  • High purity (~99.5%) suitable for research applications.
  • Soluble in DMSO; in vitro solubility approximately 100 mg/mL.
  • Storage guidance provided for powder and solutions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.