Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ro8191 | 691868-88-9 | 98.8% | 373.21 g/mol | C14H5F6N5O | 100 MG
SDP

Catalog No. 5000210628
Encompass_Preferred

RO8191 is an orally active small-molecule interferon receptor agonist that binds IFNα/β receptor and activates JAK/STAT signalling, showing antiviral activity in HCV replicon assays.

  • Orally active IFN receptor agonist that activates JAK/STAT and ISG expression.
  • Shown activity in HCV replicon assays (IC50 ≈ 200 nM) and reported anti-HBV effects.
  • Chemical formula C14H5F6N5O; molecular weight 373.21 g/mol; CAS 691868-88-9.
  • Purity reported ~98.8% (datasheet); supplier documentation also lists ≥99.0%.
  • Store under inert gas at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month) under nitrogen.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-10628 Supplier Medchemexpress LLC Supplier No. HYW063968100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RO8191 is an orally active small-molecule interferon receptor agonist that binds IFNα/β receptor and activates JAK/STAT signalling, showing antiviral activity in HCV replicon assays.

  • Orally active IFN receptor agonist that activates JAK/STAT and ISG expression.
  • Shown activity in HCV replicon assays (IC50 ≈ 200 nM) and reported anti-HBV effects.
  • Chemical formula C14H5F6N5O; molecular weight 373.21 g/mol; CAS 691868-88-9.
  • Purity reported ~98.8% (datasheet); supplier documentation also lists ≥99.0%.
  • Store under inert gas at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month) under nitrogen.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.