Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Road Hustler Premium Grain Cowhide Leather Drivers Gloves - Keystone Thumb

Catalog No. 19042130 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-042130 X-Large
19-042-124A Medium
19-042125 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-042130 Supplier MCR Safety Supplier No. 3200XL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed of unlined premium cow grain leather with a shirred elastic back, rolled leather hem, and a keystone thumb pattern.

  • Premium Grade Grain Cowhide Leather
  • Keystone Thumb
  • Shirred Elastic Back
  • Rolled Leather Hem

Specifications

ANSI Abrasion Rating ANSI 4
Color Beige
Grade Premium
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size X-Large
Style Gunn
For Use With (Application) Carpentry, Farming, Landscaping, Airport , Agriculture, Transportation, Construction, and Manufacturing
Glove Cuff Style Open
Lining Unlined
Packaging Type Polybag
Series 3200
Thumb Style Keystone
Length (English) 9.6 in.
Length (Metric) 24.4 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.