Learn More
Cayman Chemical Robenacoxib, Purity: ≥98%, CAS: 220991-32-2, 50 mg
SDP
Catalog No. 502944352
Encompass_Preferred
An NSAID and COX-2 inhibitor (Ki = 30 nM for the human enzyme); selective for COX-2 over COX-1 (Ki = 800 nM for the ovine enzyme); inhibits IL-1β-induced PGE2 production in human dermal fibroblasts (IC50 = 31 nM); reduces carrageenan-induced paw edema (ID50 = 0.4 mg/kg), has antinociceptive activity in the Randall-Selitto test, and inhibits LPS-induced fever in rats
Catalog No. 50-294-4352 Supplier Cayman Chemical Supplier No. 4221850 MG
An NSAID and COX-2 inhibitor (Ki = 30 nM for the human enzyme); selective for COX-2 over COX-1 (Ki = 800 nM for the ovine enzyme); inhibits IL-1β-induced PGE2 production in human dermal fibroblasts (IC50 = 31 nM); reduces carrageenan-induced paw edema (ID50 = 0.4 mg/kg), has antinociceptive activity in the Randall-Selitto test, and inhibits LPS-induced fever in rats