Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Robenacoxib-d5 | 2012598-81-9 | 99.6% | 332.30 | C16H8D5F4NO2 | 1 MG
SDP

Catalog No. 5000227765
Encompass_Preferred

Robenacoxib-d5 is a deuterium-labeled form of the COX inhibitor robenacoxib supplied for research use. It is intended as a tracer or internal standard in analytical and pharmacokinetic studies, and is provided with characterized molecular data and high chemical purity for reliable quantitation.

  • Deuterium-labeled tracer for quantitative analysis and pharmacokinetic studies.
  • High purity suitable for use as an internal standard.
  • Characterized molecular data: CAS 2012598-81-9, formula C16H8D5F4NO2, molecular weight 332.30.
  • Available in small, precisely measured quantities for analytical workflows.

Catalog No. 50-002-27765 Supplier Medchemexpress LLC Supplier No. HY118078S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Robenacoxib-d5 is a deuterium-labeled form of the COX inhibitor robenacoxib supplied for research use. It is intended as a tracer or internal standard in analytical and pharmacokinetic studies, and is provided with characterized molecular data and high chemical purity for reliable quantitation.

  • Deuterium-labeled tracer for quantitative analysis and pharmacokinetic studies.
  • High purity suitable for use as an internal standard.
  • Characterized molecular data: CAS 2012598-81-9, formula C16H8D5F4NO2, molecular weight 332.30.
  • Available in small, precisely measured quantities for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.