Learn More
Medchemexpress LLC Robenacoxib-d5 | 2012598-81-9 | 99.6% | 332.30 | C16H8D5F4NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY118078S5MG
Encompass_Preferred
Robenacoxib-d5 is the deuterium-labeled isotopologue of robenacoxib supplied as a stable isotope standard for analytical and research applications. It is intended primarily for use as an internal standard or tracer in quantitative mass spectrometry and method development.
  • Deuterium-labeled internal standard for accurate quantification.
  • Suitable for LC-MS method development and validation.
  • Reduces matrix effects and improves assay precision.
  • Provided in milligram quantities compatible with analytical workflows.
  • Molecular formula C16H8D5F4NO2 and molecular weight 332.30.
Catalog No. 50-002-26508