Learn More
Medchemexpress LLC Robenidine-d8 hydrochloride | 1173097-77-2 | 99.0% | 25 MG
SDP
Catalog No. 5000411968
Encompass_Preferred
Robenidine-d8 hydrochloride is the deuterium labeled version of Robenidine hydrochloride. Robenidine hydrochloride acts as an anticoccidial agent and is also effective against MRSA and VRE, with MIC50s of 8.1 and 4.7 μM, respectively.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Incorporated stable heavy isotopes into drug molecules for use as tracers in drug development.
  • Potential to influence the pharmacokinetic and metabolic profiles of drugs through deuteration.
Catalog No. 50-004-11968 Supplier Medchemexpress LLC Supplier No. 25MGHYB2157S
Robenidine-d8 hydrochloride is the deuterium labeled version of Robenidine hydrochloride. Robenidine hydrochloride acts as an anticoccidial agent and is also effective against MRSA and VRE, with MIC50s of 8.1 and 4.7 μM, respectively.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Incorporated stable heavy isotopes into drug molecules for use as tracers in drug development.
  • Potential to influence the pharmacokinetic and metabolic profiles of drugs through deuteration.