Learn More
Medchemexpress LLC HY-B2157 100mg Medchemexpress, Robenidine hydrochloride CAS:25875-50-7 Purity:>98%
SDP
Catalog No. 501933088
Encompass_Preferred
Medchemexpress, HY-B2157 100mg Robenidine hydrochloride CAS:25875-50-7 Robenidine hydrochloride is an anticoccidial agent which is also active against MRSA and VRE with MIC 50 s of 8.1 and 4.7 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3088 Supplier Medchemexpress LLC Supplier No. HYB2157100MG
Medchemexpress, HY-B2157 100mg Robenidine hydrochloride CAS:25875-50-7 Robenidine hydrochloride is an anticoccidial agent which is also active against MRSA and VRE with MIC 50 s of 8.1 and 4.7 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.