Learn More
Medchemexpress LLC HY-N1347 5mg Medchemexpress, Robinetin CAS:490-31-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN13475MG
Encompass_Preferred
Medchemexpress, HY-N1347 5mg Robinetin CAS:490-31-3 Robinetin (3,3',4',5',7-Pentahydroxyflavone), a naturally occurring flavonoid with remarkable ‘two color’ intrinsic fluorescence properties, has antifungal, antiviral, antibacterial, antimutagenesis, and antioxidant activity. Robinetin also can inhibit lipid peroxidation and protein glycosylation[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2836