Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Robo1 protein, rat (HEK293, His) | >95.0% | 100UG
SDP

Catalog No. 5000174838
Encompass_Preferred

Recombinant rat ROBO1 protein expressed in HEK293 cells with a C-terminal His tag. It functions as an extracellular receptor for SLIT1 and SLIT2, mediating cell migration and axonal guidance, and is supplied lyophilized for long-term storage and downstream biochemical or cell-based assays.

  • Recombinant rat ROBO1 expressed in HEK293 cells.
  • C-terminal His tag for affinity purification.
  • Glycosylated protein with molecular weight ≈70-95 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Endotoxin <1 EU/μg, suitable for sensitive assays.
  • Stable at -20°C unopened; reconstitute and aliquot for storage.

Catalog No. 50-001-74838 Supplier Medchemexpress LLC Supplier No. HYP702544100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat ROBO1 protein expressed in HEK293 cells with a C-terminal His tag. It functions as an extracellular receptor for SLIT1 and SLIT2, mediating cell migration and axonal guidance, and is supplied lyophilized for long-term storage and downstream biochemical or cell-based assays.

  • Recombinant rat ROBO1 expressed in HEK293 cells.
  • C-terminal His tag for affinity purification.
  • Glycosylated protein with molecular weight ≈70-95 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Endotoxin <1 EU/μg, suitable for sensitive assays.
  • Stable at -20°C unopened; reconstitute and aliquot for storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.