Learn More
Medchemexpress LLC Robo1 recombinant protein (rat, HEK293, C-terminal His tag) | 10UG
SDP
Catalog No. 5000176105
Encompass_Preferred
Recombinant rat ROBO1 receptor protein produced in HEK293 cells with a C-terminal His tag. The product is glycosylated and purified to high quality, making it suitable for binding, structural, and biochemical studies. Supplied as 10 μg with an approximate molecular weight of 70-95 kDa and >95% purity. Store at -20°C and aliquot for extended storage; after reconstitution it is stable at 4°C for short-term use.
Catalog No. 50-001-76105 Supplier Medchemexpress LLC Supplier No. HYP70254410UG
Recombinant rat ROBO1 receptor protein produced in HEK293 cells with a C-terminal His tag. The product is glycosylated and purified to high quality, making it suitable for binding, structural, and biochemical studies. Supplied as 10 μg with an approximate molecular weight of 70-95 kDa and >95% purity. Store at -20°C and aliquot for extended storage; after reconstitution it is stable at 4°C for short-term use.