Learn More
Aviva Systems Biology Corp ROBO2 Antibody - N-terminal region (ARP45396_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45396_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ROBO2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ROBO2. Alternate names: KIAA1568, SAX3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59019