Learn More
Biomatik Corporation Robo2 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10899
Encompass_Preferred
Robo2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ROBO2. The Robo2 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Roundabout Homolog 2 Protein (450-660Aa). Applications: ELISA, WB. Target Synonyms: ROBO2. Accession Number: Q9HCK4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9258