Learn More
STEMCELL Technologies RoboSep™ CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974681
Encompass
The EasySep™ Human CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kit is designed to isolate CD4+CD127lowCD49d- cells from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells, CD127high, CD49d+ cells and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube.
Catalog No. 50-197-4681 Supplier STEMCELL Technologies Supplier No. 19232RF
The EasySep™ Human CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kit is designed to isolate CD4+CD127lowCD49d- cells from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells, CD127high, CD49d+ cells and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube.