Learn More
STEMCELL Technologies RoboSep™ FITC Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974552
Encompass
The EasySep™ FITC Positive Selection Kit II is designed to isolate cells that are labeled with FITC-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing FITC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ FITC Positive Selection Kit (Catalog #18558) for even faster cell isolations.
Catalog No. 50-197-4552 Supplier STEMCELL Technologies Supplier No. 17682RF
The EasySep™ FITC Positive Selection Kit II is designed to isolate cells that are labeled with FITC-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing FITC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ FITC Positive Selection Kit (Catalog #18558) for even faster cell isolations.