Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit, For processing: 60 mL buffy coat or whole blood processed
SDP
Catalog No. 501974577
Encompass
The EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit is designed to isolate CD19+ cells from whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is present on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.This product replaces the EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084) for even faster cell isolations.
Catalog No. 50-197-4577 Supplier STEMCELL Technologies Supplier No. 17874RF
The EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit is designed to isolate CD19+ cells from whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is present on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.This product replaces the EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084) for even faster cell isolations.