Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Whole Blood B Cell Positive Selection Kit, For processing: 60 mL buffy coat or whole blood processed
SDP
Catalog No. 501974595
Encompass
The EasySep™ HLA Chimerism Whole Blood B Cell Positive Selection Kit is designed to isolate B cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19, CD20 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is present on mature B cells and B cell progenitors, but is lost upon maturation to plasma cells. The antigen CD20 is expressed on all stages of B cell development except on pro B cells or plasma cells.This product replaces the EasySep™ HLA Whole Blood B Cell Positive Selection Kit (Catalog #18184HLA) for even faster cell isolations.
Catalog No. 50-197-4595 Supplier STEMCELL Technologies Supplier No. 17886RF
The EasySep™ HLA Chimerism Whole Blood B Cell Positive Selection Kit is designed to isolate B cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19, CD20 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is present on mature B cells and B cell progenitors, but is lost upon maturation to plasma cells. The antigen CD20 is expressed on all stages of B cell development except on pro B cells or plasma cells.This product replaces the EasySep™ HLA Whole Blood B Cell Positive Selection Kit (Catalog #18184HLA) for even faster cell isolations.