Learn More
STEMCELL Technologies RoboSep™ HLA T Cell Enrichment Kit with Filter Tips, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974669
Encompass
The EasySep™ HLA T Cell Enrichment Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-T cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube and are ready for serology or flow cytometry crossmatch assays.
Catalog No. 50-197-4669 Supplier STEMCELL Technologies Supplier No. 19051HLARF
The EasySep™ HLA T Cell Enrichment Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-T cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube and are ready for serology or flow cytometry crossmatch assays.