Learn More
STEMCELL Technologies RoboSep™ HLA Total Lymphocyte Enrichment: Complete Processing Kit for Whole Blood with Filter Tips, For processing: 200 mL whole blood processed
SDP
Catalog No. 501974737
Encompass
The EasySep™ Total Lymphocyte Enrichment Kit: Complete Processing Kit for Whole Blood is designed to isolate human lymphocytes from HetaSep™-treated whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD14, CD15, CD16, CD33, CD36, CD41, CD66b, glycophorin A and dextran-coated magnetic particles. The labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. 50-197-4737 Supplier STEMCELL Technologies Supplier No. 19961HLARF
The EasySep™ Total Lymphocyte Enrichment Kit: Complete Processing Kit for Whole Blood is designed to isolate human lymphocytes from HetaSep™-treated whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD14, CD15, CD16, CD33, CD36, CD41, CD66b, glycophorin A and dextran-coated magnetic particles. The labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.