Learn More
STEMCELL Technologies RoboSep™ HLA Whole Blood T Cell Enrichment Kit with Filter Tips, For processing: 200 mL whole blood processed
SDP
Catalog No. 501974733
Encompass
The Complete Processing Kit for whole blood uses EasySep™ HLA T Cell Enrichment Kit to isolate human T cells from HetaSep™-treated whole blood by negative selection. Unwanted cells are targeted for removal with tetrameric antibody complexes recognizing non-T cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. 50-197-4733 Supplier STEMCELL Technologies Supplier No. 19951HLARF
The Complete Processing Kit for whole blood uses EasySep™ HLA T Cell Enrichment Kit to isolate human T cells from HetaSep™-treated whole blood by negative selection. Unwanted cells are targeted for removal with tetrameric antibody complexes recognizing non-T cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.