Learn More
STEMCELL Technologies RoboSep™ Human B Cell Enrichment Kit with Filter Tips, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974673
Encompass
The EasySep™ Human B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For even faster cell isolations, we recommend the new EasySep™ Human B Cell Isolation Kit (17954) (/products/easysep-human-b-cell-isolation-kit.html), which isolates cells in just 9 minutes.
Catalog No. 50-197-4673 Supplier STEMCELL Technologies Supplier No. 19054RF
The EasySep™ Human B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For even faster cell isolations, we recommend the new EasySep™ Human B Cell Isolation Kit (17954) (/products/easysep-human-b-cell-isolation-kit.html), which isolates cells in just 9 minutes.