Learn More
STEMCELL Technologies RoboSep™ Human Basophil Isolation Kit, For isolating: 1 x 10^9 cells
SDP
Catalog No. 501974623
Encompass
The EasySep™ Human Basophil Isolation Kit is designed to isolate basophils from a polymorphonuclear cell-rich fraction by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-basophils and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation of fresh peripheral blood.This product replaces the EasySep™ Human Basophil Enrichment Kit (Catalog #19069) for even faster cell isolations.
Catalog No. 50-197-4623 Supplier STEMCELL Technologies Supplier No. 17969RF
The EasySep™ Human Basophil Isolation Kit is designed to isolate basophils from a polymorphonuclear cell-rich fraction by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-basophils and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation of fresh peripheral blood.This product replaces the EasySep™ Human Basophil Enrichment Kit (Catalog #19069) for even faster cell isolations.