Learn More
STEMCELL Technologies RoboSep Human CD19 Positive Selection Kit II, For processing: 1 x 10 9 cells
SDP
Catalog No. 502390099
Encompass
The EasySep Human CD19 Positive Selection Kit II is designed to isolate CD19 cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.
Catalog No. 50-239-0099 Supplier STEMCELL Technologies Supplier No. 17854RF
The EasySep Human CD19 Positive Selection Kit II is designed to isolate CD19 cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.