Learn More
STEMCELL Technologies RoboSep™ Human Eosinophil Isolation Kit, For isolating: 1 x 10^9 cells
SDP
Catalog No. 501974614
Encompass
The EasySep™ Human Eosinophil Isolation Kit is designed to isolate eosinophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-eosinophils and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The kit is compatible with cells prepared using either Lymphoprep™ (Catalog #07801) centrifugation (preferred for higher cell purities) or HetaSep™ (Catalog #07906) sedimentation (preferred for faster, lysis-free sample processing).This product replaces the EasySep™ Human Eosinophil Enrichment Kit (Catalog #19256) for even faster cell isolations.
Catalog No. 50-197-4614 Supplier STEMCELL Technologies Supplier No. 17956RF
The EasySep™ Human Eosinophil Isolation Kit is designed to isolate eosinophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-eosinophils and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The kit is compatible with cells prepared using either Lymphoprep™ (Catalog #07801) centrifugation (preferred for higher cell purities) or HetaSep™ (Catalog #07906) sedimentation (preferred for faster, lysis-free sample processing).This product replaces the EasySep™ Human Eosinophil Enrichment Kit (Catalog #19256) for even faster cell isolations.