Learn More
STEMCELL Technologies RoboSep™ Human Memory CD4 T Cell Enrichment Kit with Filter Tips, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974679
Encompass
The EasySep™ Human Memory CD4 T Cell Enrichment Kit is designed to isolate memory CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD8, CD14, CD16, CD19, CD20, CD36, CD45RA, CD56, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. 50-197-4679 Supplier STEMCELL Technologies Supplier No. 19157RF
The EasySep™ Human Memory CD4 T Cell Enrichment Kit is designed to isolate memory CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD8, CD14, CD16, CD19, CD20, CD36, CD45RA, CD56, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.