Learn More
STEMCELL Technologies RoboSep™ Human Memory CD8+ T Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974680
Encompass
The EasySep™ Human Memory CD8+ T Cell Enrichment Kit is designed to isolate memory CD8+ T cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD4, CD14, CD16, CD19, CD20, CD34, CD36, CD45RA, CD56, CD61, CD66b, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. 50-197-4680 Supplier STEMCELL Technologies Supplier No. 19159RF
The EasySep™ Human Memory CD8+ T Cell Enrichment Kit is designed to isolate memory CD8+ T cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD4, CD14, CD16, CD19, CD20, CD34, CD36, CD45RA, CD56, CD61, CD66b, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.