Learn More
STEMCELL Technologies RoboSep Human Naïve B Cell Isolation Kit, For processing: 1 x 10 9 cells
SDP
Catalog No. 502390096
Encompass
The EasySep Human Naïve B Cell Isolation Kit is designed to isolate naïve B cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep magnet and simply pouring or pipetting the desired cells into a new tube. This product can be used in place of the EasySep Human Naïve B Cell Enrichment Kit (Catalog 19254) for even faster cell isolations.
Catalog No. 50-239-0096 Supplier STEMCELL Technologies Supplier No. 17254RF
The EasySep Human Naïve B Cell Isolation Kit is designed to isolate naïve B cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep magnet and simply pouring or pipetting the desired cells into a new tube. This product can be used in place of the EasySep Human Naïve B Cell Enrichment Kit (Catalog 19254) for even faster cell isolations.