Learn More
STEMCELL Technologies RoboSep™ Human Naïve CD4+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974691
Encompass
The EasySep™ Human Naïve CD4+ T Cell Isolation Kit is designed to isolate naive CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes and dextran-coated magnetic particles. CD45RO+ cells are targeted for removal with a biotinylated anti-CD45RO antibody, and a bispecific Tetrameric Antibody Complex that recognizes biotin and dextran. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the new EasySep™ Human Naïve CD4+ T Cell Isolation Kit II (17555) (https://www.stemcell.com/products/easysep-human-naive-cd4-t-cell-isolation-kit-ii.html) which isolates cells in just 11 minutes.
Catalog No. 50-197-4691 Supplier STEMCELL Technologies Supplier No. 19555RF
The EasySep™ Human Naïve CD4+ T Cell Isolation Kit is designed to isolate naive CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes and dextran-coated magnetic particles. CD45RO+ cells are targeted for removal with a biotinylated anti-CD45RO antibody, and a bispecific Tetrameric Antibody Complex that recognizes biotin and dextran. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the new EasySep™ Human Naïve CD4+ T Cell Isolation Kit II (17555) (https://www.stemcell.com/products/easysep-human-naive-cd4-t-cell-isolation-kit-ii.html) which isolates cells in just 11 minutes.