Learn More
STEMCELL Technologies RoboSep™ Human Naïve CD4+ T Cell Isolation Kit II, Size: 1 x 10^9 cells
SDP
Catalog No. 501974541
Encompass
The EasySep™ Human Naïve CD4+ T Cell Isolation Kit II is designed to isolate naïve CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.This kit replaces the EasySep™ Human Naïve CD4+ T Cell Enrichment Kit (Catalog #19155) and EasySep™ Human Naïve CD4+ T Cell Isolation Kit (Catalog #19555) for even faster cell isolations.
Catalog No. 50-197-4541 Supplier STEMCELL Technologies Supplier No. 17555RF
The EasySep™ Human Naïve CD4+ T Cell Isolation Kit II is designed to isolate naïve CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.This kit replaces the EasySep™ Human Naïve CD4+ T Cell Enrichment Kit (Catalog #19155) and EasySep™ Human Naïve CD4+ T Cell Isolation Kit (Catalog #19555) for even faster cell isolations.