Learn More
STEMCELL Technologies RoboSep™ Human NK Cell Enrichment Kit with Filter Tips, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974674
Encompass
The EasySep™ Human NK Cell Enrichment Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-NK cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the new EasySep™ Human NK Cell Isolation Kit (17955) (/products/easysep-human-nk-cell-isolation-kit.html) which isolates cells in just 8 minutes.
Catalog No. 50-197-4674 Supplier STEMCELL Technologies Supplier No. 19055RF
The EasySep™ Human NK Cell Enrichment Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-NK cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the new EasySep™ Human NK Cell Isolation Kit (17955) (/products/easysep-human-nk-cell-isolation-kit.html) which isolates cells in just 8 minutes.