Learn More
STEMCELL Technologies RoboSep™ Human Resting CD4+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974618
Encompass
The EasySep™ Human Resting CD4+ T Cell Isolation Kit is designed to isolate resting CD4+ T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Catalog No. 50-197-4618 Supplier STEMCELL Technologies Supplier No. 17962RF
The EasySep™ Human Resting CD4+ T Cell Isolation Kit is designed to isolate resting CD4+ T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.