Learn More
STEMCELL Technologies RoboSep Human T Cell Enrichment Kit with Filter Tips, For processing: 1 x 10 9 cells
SDP
Catalog No. 502390116
Encompass
The EasySep Human T Cell Enrichment Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-T cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. 50-239-0116 Supplier STEMCELL Technologies Supplier No. 19051RF
The EasySep Human T Cell Enrichment Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-T cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep magnet without the use of columns. Desired cells are poured off into a new tube.