Learn More
STEMCELL Technologies RoboSep™ Mouse Biotin Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974547
Encompass
The EasySep™ Mouse Biotin Positive Selection Kit II is designed to isolate cells that are labeled with biotinylated antibodies. Desired cells are targeted with antibody complexes recognizing biotin and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Mouse Biotin Positive Selection Kit (Catalog #18556). For highly purified particle-free cells, we recommend the EasySep™ Release Mouse Biotin Positive Selection Kit (Catalog #17655). (https://www.stemcell.com/products/easysep-release-mouse-biotin-positive-selection-kit.html)
Catalog No. 50-197-4547 Supplier STEMCELL Technologies Supplier No. 17665RF
The EasySep™ Mouse Biotin Positive Selection Kit II is designed to isolate cells that are labeled with biotinylated antibodies. Desired cells are targeted with antibody complexes recognizing biotin and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Mouse Biotin Positive Selection Kit (Catalog #18556). For highly purified particle-free cells, we recommend the EasySep™ Release Mouse Biotin Positive Selection Kit (Catalog #17655). (https://www.stemcell.com/products/easysep-release-mouse-biotin-positive-selection-kit.html)