Learn More
STEMCELL Technologies RoboSep™ Mouse CD25 Regulatory T cell Positive Selection Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974654
Encompass
The EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kit is designed to isolate highly purified CD25+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. The kit targets CD25 regulatory T cells for positive selection with a PE-labeled antibody directed against the CD25 surface marker. Desired cells are labeled with antibodies and magnetic particles. The cells are separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube.
Catalog No. 50-197-4654 Supplier STEMCELL Technologies Supplier No. 18782RF
The EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kit is designed to isolate highly purified CD25+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. The kit targets CD25 regulatory T cells for positive selection with a PE-labeled antibody directed against the CD25 surface marker. Desired cells are labeled with antibodies and magnetic particles. The cells are separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube.