Learn More
STEMCELL Technologies RoboSep™ Mouse CD8+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974725
Encompass
The EasySep™ Mouse CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-CD8+ T cells and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.This product replaces the EasySep™ Mouse CD8+ T Cell Enrichment Kit (Catalog #19753) for even faster cell isolations.
Catalog No. 50-197-4725 Supplier STEMCELL Technologies Supplier No. 19853RF
The EasySep™ Mouse CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-CD8+ T cells and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.This product replaces the EasySep™ Mouse CD8+ T Cell Enrichment Kit (Catalog #19753) for even faster cell isolations.