Learn More
STEMCELL Technologies RoboSep™ Mouse CD90.2 Positive Selection Kit II, For processing: 2 x 10^9 cells
SDP
Catalog No. 501974657
Encompass
The EasySep™ Mouse CD90.2 Positive Selection Kit II is designed to isolate CD90.2+ (Thy1.2+) cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognizing CD90.2 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Mouse CD90.2 Positive Selection Kit (Catalog #18751) for even faster cell isolations and does not result in the labeling of isolated cells with PE.
Catalog No. 50-197-4657 Supplier STEMCELL Technologies Supplier No. 18951RF
The EasySep™ Mouse CD90.2 Positive Selection Kit II is designed to isolate CD90.2+ (Thy1.2+) cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognizing CD90.2 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Mouse CD90.2 Positive Selection Kit (Catalog #18751) for even faster cell isolations and does not result in the labeling of isolated cells with PE.