Learn More
STEMCELL Technologies RoboSep™ Mouse FITC Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974550
Encompass
The EasySep™ Mouse FITC Positive Selection Kit II is designed to isolate mouse cells that are labeled with FITC-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing FITC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Mouse FITC Positive Selection Kit (Catalog #18555).
Catalog No. 50-197-4550 Supplier STEMCELL Technologies Supplier No. 17668RF
The EasySep™ Mouse FITC Positive Selection Kit II is designed to isolate mouse cells that are labeled with FITC-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing FITC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Mouse FITC Positive Selection Kit (Catalog #18555).