Learn More
STEMCELL Technologies EasySep™ Mouse Pan-Nave T Cell Isolation Kit, 1 x 10^9 cells
SDP
Catalog No. 501974723
Encompass
The EasySep™ Mouse Pan-B Cell Isolation Kit is designed to isolate all B cells, including conventional B-2 B cells, B-1 B cells and plasma cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non pan-B cells (CD4, CD8, CD11c, CD49b, CD90.2, Ly-6C/G (Gr-1), TER119) and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. This kit is compatible for use with cells from disease models where the malignant cells (B-CLL) express CD43 or CD11b. For isolation of conventional B cells only, we recommend using the EasySep™ Mouse B Cell Isolation Kit (Catalog #19854).
Catalog No. 50-197-4723 Supplier STEMCELL Technologies Supplier No. 19844RF
The EasySep™ Mouse Pan-B Cell Isolation Kit is designed to isolate all B cells, including conventional B-2 B cells, B-1 B cells and plasma cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non pan-B cells (CD4, CD8, CD11c, CD49b, CD90.2, Ly-6C/G (Gr-1), TER119) and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. This kit is compatible for use with cells from disease models where the malignant cells (B-CLL) express CD43 or CD11b. For isolation of conventional B cells only, we recommend using the EasySep™ Mouse B Cell Isolation Kit (Catalog #19854).