Learn More
STEMCELL Technologies RoboSep™ Rat CD4+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1530640
Encompass
The EasySep™ Rat CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Catalog No. NC1530640 Supplier STEMCELL Technologies Supplier No. 19642RF
The EasySep™ Rat CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.